2-[(3-Chlorobenzyl)oxy]-3-methoxybenzaldehyde
Supplier: Matrix Scientific
028512-500MG
101826-286EA
146.15
USD
101826-286
2-[(3-Chlorobenzyl)oxy]-3-methoxybenzaldehyde
2-[(3-Chlorobenzyl)oxy]-3-methoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₅H₁₃ClO₃ |
MDL Number:
MFCD03422406 CAS Number: 588678-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...