2-[(2-Chlorobenzyl)oxy]-3-methoxybenzaldehyde

Supplier: Matrix Scientific

028510-500MG
101826-282EA 146.15 USD
101826-282
2-[(2-Chlorobenzyl)oxy]-3-methoxybenzaldehyde
2-[(2-Chlorobenzyl)oxy]-3-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₃ClO₃
MDL Number: MFCD03422404
CAS Number: 52803-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR