2-[(2-Fluorobenzyl)oxy]-3-methoxybenzaldehyde

Supplier: Matrix Scientific

028508-500MG
101826-278EA 146.15 USD
101826-278
2-[(2-Fluorobenzyl)oxy]-3-methoxybenzaldehyde
2-[(2-Fluorobenzyl)oxy]-3-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₃FO₃
MDL Number: MFCD03422402
CAS Number: 588687-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR