2-[(4-Chlorobenzyl)oxy]-3-methoxybenzaldehyde

Supplier: Matrix Scientific

028506-500MG
101826-274EA 109.25 USD
101826-274
2-[(4-Chlorobenzyl)oxy]-3-methoxybenzaldehyde
2-[(4-Chlorobenzyl)oxy]-3-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₃ClO₃
MDL Number: MFCD03422401
CAS Number: 52803-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR