2-[(3-Methoxybenzyl)oxy]-3-methoxybenzaldehyde

Supplier: Matrix Scientific

028502-500MG
101826-266EA 86.15 USD
101826-266
2-[(3-Methoxybenzyl)oxy]-3-methoxybenzaldehyde
2-[(3-Methoxybenzyl)oxy]-3-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MDL Number: MFCD00553744
CAS Number: 350693-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR