4-(Allyloxy)benzaldehyde

Supplier: Matrix Scientific

028466-500MG
101826-194EA 15.38 USD
101826-194
4-(Allyloxy)benzaldehyde
4-(Allyloxy)benzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MW: 162.19
Boiling Pt: 142 °C (10 Torr)
Melting Pt: 179 °C
MDL Number: MFCD00014133
CAS Number: 40663-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR