3-Isobutoxybenzaldehyde

Supplier: Matrix Scientific

028450-500MG
101826-160EA 267.69 USD
101826-160
3-Isobutoxybenzaldehyde
3-Isobutoxybenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD01993681
CAS Number: 67698-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR