3-Iodo-2-(4-morpholino)pyridine

Supplier: Matrix Scientific

Synonyms: 4-(3-Iodo-2-pyridinyl-morpholine

026100-500MG
101821-606EA 253.85 USD
101821-606
3-Iodo-2-(4-morpholino)pyridine
3-Iodo-2-(4-morpholino)pyridine

Hazard: Irritant

Formula: C₉H₁₁IN₂O
MDL Number: MFCD04039864
CAS Number: 470463-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR