N-Acetyl-DL-phenylalanine

Supplier: Matrix Scientific

Synonyms: Ac-DL-Phe-OH

025960-500MG
101821-328EA 461.54 USD
101821-328
N-Acetyl-DL-phenylalanine
N-Acetyl-DL-phenylalanine

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MW: 207.229 g/mol
MDL Number: MFCD00063157
CAS Number: 2901-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR