N-phenylanthranilic acid

Supplier: Matrix Scientific

Synonyms: 2-(Phenylamino)benzoic acid

025959-500MG
101821-326EA 306.15 USD
101821-326
N-phenylanthranilic acid
N-phenylanthranilic acid

Hazard: Irritant

Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
MDL Number: MFCD00002421
CAS Number: 91-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR