(4-Nitrophenoxy)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(4-Nitrophenoxy)acetic acid4-Nitrophenoxyacetic acid

025952-500MG
101821-310EA 306.15 USD
101821-310
(4-Nitrophenoxy)acetic acid
(4-Nitrophenoxy)acetic acid

Hazard: Irritant

Formula: (C₆H₄(NO₂))OCH₂COOH
MW: 197.15 g/mol
MDL Number: MFCD00017030
CAS Number: 1798-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR