(4-Tolyloxy)acetic acid

Supplier: Matrix Scientific

Synonyms: (4-Methylphenoxy)acetic acid4-Methylphenoxyacetic acid2-(p-Tolyloxy)acetic acid

025951-500MG
101821-308EA 306.15 USD
101821-308
(4-Tolyloxy)acetic acid
(4-Tolyloxy)acetic acid

Hazard: Irritant

Formula: (C₆H₄(CH₃))OCH₂COOH
MW: 166.18 g/mol
MDL Number: MFCD00014365
CAS Number: 940-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR