2-Amino-4,5-dimethylthiophene-3-carboxamide

Supplier: Matrix Scientific

025949-500MG
101821-302EA 95.38 USD
101821-302
2-Amino-4,5-dimethylthiophene-3-carboxamide
2-Amino-4,5-dimethylthiophene-3-carboxamide

Hazard: Irritant

Formula: C₇H₁₀N₂OS
MW: 170.23 g/mol
MDL Number: MFCD00617251
CAS Number: 51486-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR