4-Amino-N-(2,6-dimethylphenyl)benzamide

Supplier: Matrix Scientific

025843-500MG
101821-088EA 306.15 USD
101821-088
4-Amino-N-(2,6-dimethylphenyl)benzamide
4-Amino-N-(2,6-dimethylphenyl)benzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MW: 240.3 g/mol
MDL Number: MFCD00865288
CAS Number: 787-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR