4-Amino-N-propylbenzamide

Supplier: Matrix Scientific

025839-500MG
101821-080EA 160 USD
101821-080
4-Amino-N-propylbenzamide
4-Amino-N-propylbenzamide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O
MDL Number: MFCD01940421
CAS Number: 38681-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR