4-Amino-N-(2,3-dimethylphenyl)benzamide

Supplier: Matrix Scientific

025832-500MG
101821-066EA 306.15 USD
101821-066
4-Amino-N-(2,3-dimethylphenyl)benzamide
4-Amino-N-(2,3-dimethylphenyl)benzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD01757449
CAS Number: 29027-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR