4-Amino-N-benzylbenzamide

Supplier: Matrix Scientific

025818-500MG
101821-038EA 306.15 USD
101821-038
4-Amino-N-benzylbenzamide
4-Amino-N-benzylbenzamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD00440145
CAS Number: 54977-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR