4-Amino-N-(3,5-dimethylphenyl)benzamide

Supplier: Matrix Scientific

025813-500MG
101821-028EA 228.85 USD
101821-028
4-Amino-N-(3,5-dimethylphenyl)benzamide
4-Amino-N-(3,5-dimethylphenyl)benzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD03166484
CAS Number: 97042-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR