3-Amino-N-(2,3-dimethylphenyl)benzamide

Supplier: Matrix Scientific

025763-500MG
101820-904EA 306.15 USD
101820-904
3-Amino-N-(2,3-dimethylphenyl)benzamide
3-Amino-N-(2,3-dimethylphenyl)benzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD03070710
CAS Number: 102630-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR