3-Amino-N-benzylbenzamide

Supplier: Matrix Scientific

025747-500MG
101820-872EA 309.23 USD
101820-872
3-Amino-N-benzylbenzamide
3-Amino-N-benzylbenzamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD00523631
CAS Number: 54977-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR