3-Amino-N-(4-methylphenyl)benzamide

Supplier: Matrix Scientific

025746-500MG
101820-870EA 306.15 USD
101820-870
3-Amino-N-(4-methylphenyl)benzamide
3-Amino-N-(4-methylphenyl)benzamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD00443925
CAS Number: 14315-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR