3-Amino-N-(3,4-dimethylphenyl)benzamide

Supplier: Matrix Scientific

025729-500MG
101820-836EA 306.15 USD
101820-836
3-Amino-N-(3,4-dimethylphenyl)benzamide
3-Amino-N-(3,4-dimethylphenyl)benzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD01138443
CAS Number: 102630-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR