4-Amino-1-boc-piperidine-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 4-Amino-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid

018362-1G
101806-054EA 67.69 USD
101806-054
4-Amino-1-boc-piperidine-4-carboxylic acid
4-Amino-1-Boc-piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₄
MW: 244.29 g/mol
MDL Number: MFCD01318728
CAS Number: 183673-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR