3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine ≥97%
Supplier: Matrix Scientific
018282-25G
101805-814EA
71.3
USD
101805-814
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine ≥97%
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₂ClF₄N MW: 199.53 g/mol |
MDL Number:
MFCD00191918 CAS Number: 72537-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...