2-Ethoxy-3-iodopyridine

Supplier: Matrix Scientific

018145-1G
101805-508EA 304.62 USD
101805-508
2-Ethoxy-3-iodopyridine
2-Ethoxy-3-iodopyridine

Hazard: Irritant

Formula: C₇H₈INO
MW: 249.0490 g/mol
MDL Number: MFCD00234318
CAS Number: 766557-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR