4-Benzyloxyaniline ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Benzyloxy)aniline

017128-25G
101805-444EA 136.92 USD
101805-444
4-Benzyloxyaniline ≥97%
4-Benzyloxyaniline

Hazard: Irritant

Formula: C₁₃H₁₃NO
MW: 199.2484 g/mol
MDL Number: MFCD00025318
CAS Number: 6373-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR