3-Amino-N,N-dimethylbenzenesulfonamide

Supplier: Matrix Scientific

016779-500MG
101805-346EA 58.65 USD
101805-346
3-Amino-N,N-dimethylbenzenesulfonamide
3-Amino-N,N-dimethylbenzenesulphonamide

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MW: 200.2600 g/mol
MDL Number: MFCD00602451
CAS Number: 6274-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR