1-(3-Bromopropoxy)-3-chlorobenzene

Supplier: Matrix Scientific

016399-500MG
101804-654EA 283.08 USD
101804-654
1-(3-Bromopropoxy)-3-chlorobenzene
1-(3-Bromopropoxy)-3-chlorobenzene

Hazard: Irritant

Formula: C₉H₁₀BrClO
Boiling Pt: 115…118 °C (3 Torr)
MDL Number: MFCD00596672
CAS Number: 37142-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR