1-(3-Bromopropoxy)-4-fluorobenzene
Supplier: Matrix Scientific
016385-500MG
101804-632EA
283.08
USD
101804-632
1-(3-Bromopropoxy)-4-fluorobenzene
1-(3-Bromopropoxy)-4-fluorobenzene
Hazard: Irritant
Formula:
C₉H₁₀BrFO Boiling Pt: 134…136 °C (17 Torr) |
CAS Number:
1129-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...