4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde

Supplier: Matrix Scientific

016275-500MG
101804-426EA 283.08 USD
101804-426
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₂N₂O
MDL Number: MFCD06800625
CAS Number: 934570-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR