3-(1H-Pyrazol-1-yl)benzaldehyde

Supplier: Matrix Scientific

016271-500MG
101804-418EA 58.46 USD
101804-418
3-(1H-Pyrazol-1-yl)benzaldehyde
3-(1H-Pyrazol-1-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₀H₈N₂O
MW: 172.18 g/mol
MDL Number: MFCD06740321
CAS Number: 852227-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR