N,N-Dimethyl-N'-(5-methylfuran-2-ylmethyl)propane-1,3-diamine

Supplier: Matrix Scientific

011000-500MG
101799-632EA 267.69 USD
101799-632
N,N-Dimethyl-N'-(5-methylfuran-2-ylmethyl)propane-1,3-diamine
N,N-Dimethyl-N'-(5-methylfuran-2-ylmethyl)propane-1,3-diamine

Hazard: Irritant

Formula: C₁₁H₂₀N₂O
MW: 196.29 g/mol
MDL Number: MFCD03856658
CAS Number: 626216-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR