[1-(4-Methoxyphenyl)ethyl]pyridin-3-ylmethylamine

Supplier: Matrix Scientific

010973-500MG
101799-578EA 267.69 USD
101799-578
[1-(4-Methoxyphenyl)ethyl]pyridin-3-ylmethylamine
[1-(4-Methoxyphenyl)ethyl]pyridin-3-ylmethylamine

Hazard: Irritant

Formula: C₁₅H₁₈N₂O
MDL Number: MFCD03856636
CAS Number: 626216-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR