4-(4-Methoxyphenyl)-1,3-thiazol-2-amine

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-(4-methoxyphenyl)-1,3-thiazole4-(4-Methoxyphenyl)thiazol-2-amine

010942-500MG
101799-516EA 235.38 USD
101799-516
4-(4-Methoxyphenyl)-1,3-thiazol-2-amine
4-(4-Methoxyphenyl)-1,3-thiazol-2-amine

Hazard: Irritant

Formula: C₁₀H₁₀N₂OS
MW: 206.27 g/mol
Melting Pt: 204…205 °C
MDL Number: MFCD00170663
CAS Number: 2104-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR