1-(2-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

010875-500MG
101799-386EA 50.77 USD
101799-386
1-(2-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(2-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
Melting Pt: 165 °C
MDL Number: MFCD03724918
CAS Number: 39629-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR