N-(4-Piperidin-1-ylmethylphenyl)succinamic acid

Supplier: Matrix Scientific

010867-500MG
101799-368EA 267.69 USD
101799-368
N-(4-Piperidin-1-ylmethylphenyl)succinamic acid
N-(4-Piperidin-1-ylmethylphenyl)succinamic acid

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₃
MW: 290.37 g/mol
MDL Number: MFCD03724817
CAS Number: 510764-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR