N-(4-Pyrrolidin-1-ylmethylphenyl)succinamic acid

Supplier: Matrix Scientific

010866-500MG
101799-366EA 267.69 USD
101799-366
N-(4-Pyrrolidin-1-ylmethylphenyl)succinamic acid
N-(4-Pyrrolidin-1-ylmethylphenyl)succinamic acid

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₃
MW: 276.34 g/mol
MDL Number: MFCD03724816
CAS Number: 510764-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR