C-(2,3-Dihydrobenzofuran-2-yl)methylamine hydrochloride

Supplier: Matrix Scientific

Synonyms: 2-(Aminomethyl)-2,3-dihydrobenzo[b]furan hydrochloride

010859-500MG
101799-352EA 267.69 USD
101799-352
C-(2,3-Dihydrobenzofuran-2-yl)methylamine hydrochloride
C-(2,3-Dihydrobenzofuran-2-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClNO
MW: 185.66 g/mol
MDL Number: MFCD01659957
CAS Number: 19997-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR