(3,5-Dimethylpyrazol-1-yl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(3,5-Dimethyl-1H-pyrazol-1-yl)acetic acid

010857-500MG
101799-348EA 49.23 USD
101799-348
(3,5-Dimethylpyrazol-1-yl)acetic acid
(3,5-Dimethylpyrazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
MDL Number: MFCD00297249
CAS Number: 16034-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR