Cyclopropylpyridin-2-ylmethylamine

Supplier: Matrix Scientific

010851-500MG
101799-336EA 147.69 USD
101799-336
Cyclopropylpyridin-2-ylmethylamine
Cyclopropylpyridin-2-ylmethylamine

Hazard: Irritant

Formula: C₉H₁₂N₂
MDL Number: MFCD03821868
CAS Number: 626210-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR