(2-Chlorobenzyl)(3-imidazol-1-ylpropyl)amine

Supplier: Matrix Scientific

010811-500MG
101799-258EA 267.69 USD
101799-258
(2-Chlorobenzyl)(3-imidazol-1-ylpropyl)amine
(2-Chlorobenzyl)(3-imidazol-1-ylpropyl)amine

Hazard: Irritant

Formula: C₁₃H₁₆ClN₃
MDL Number: MFCD03821866
CAS Number: 626209-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR