4-Pyrrolidin-1-ylmethylphenylamine
Supplier: Matrix Scientific
010810-500MG
101799-256EA
189.23
USD
101799-256
4-Pyrrolidin-1-ylmethylphenylamine
4-Pyrrolidin-1-ylmethylphenylamine
Hazard: Irritant
Formula:
C₁₁H₁₆N₂ MW: 176.26 g/mol Melting Pt: 48…51 °C |
MDL Number:
MFCD03724739 CAS Number: 142335-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...