(2-Chlorobenzyl)(1-methylpiperidin-4-yl)amine

Supplier: Matrix Scientific

010793-500MG
101799-222EA 267.69 USD
101799-222
(2-Chlorobenzyl)(1-methylpiperidin-4-yl)amine
(2-Chlorobenzyl)(1-methylpiperidin-4-yl)amine

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂
MDL Number: MFCD01467751
CAS Number: 415970-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR