1-(4-Methylbenzoyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

010732-500MG
101799-100EA 267.69 USD
101799-100
1-(4-Methylbenzoyl)piperidine-4-carboxylic acid
1-(4-Methylbenzoyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MDL Number: MFCD03015384
CAS Number: 401581-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR