(2-Chlorobenzyl)(tetrahydrofuran-2-ylmethyl)amine

Supplier: Matrix Scientific

010681-500MG
101799-000EA 267.69 USD
101799-000
(2-Chlorobenzyl)(tetrahydrofuran-2-ylmethyl)amine
(2-Chlorobenzyl)(tetrahydrofuran-2-ylmethyl)amine

Hazard: Irritant

Formula: C₁₂H₁₆ClNO
MDL Number: MFCD03210836
CAS Number: 356532-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR