(2-Chlorobenzyl)pyridin-3-ylmethylamine

Supplier: Matrix Scientific

010601-500MG
101798-838EA 267.69 USD
101798-838
(2-Chlorobenzyl)pyridin-3-ylmethylamine
(2-Chlorobenzyl)pyridin-3-ylmethylamine

Hazard: Irritant

Formula: C₁₃H₁₃ClN₂
MDL Number: MFCD02609563
CAS Number: 436088-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR