Benzylpyridin-3-ylmethylamine

Supplier: Matrix Scientific

010600-500MG
101798-836EA 92.31 USD
101798-836
Benzylpyridin-3-ylmethylamine
Benzylpyridin-3-ylmethylamine

Hazard: Irritant

Formula: C₁₃H₁₄N₂
MW: 198.27 g/mol
MDL Number: MFCD00716866
CAS Number: 63361-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR