4-Ethyl-5-furan-2-yl-4H-[1,2,4]triazole-3-thiol

Supplier: Matrix Scientific

010585-500MG
101798-806EA 267.69 USD
101798-806
4-Ethyl-5-furan-2-yl-4H-[1,2,4]triazole-3-thiol
4-Ethyl-5-furan-2-yl-4H-[1,2,4]triazole-3-thiol

Hazard: Irritant

Formula: C₈H₉N₃OS
MDL Number: MFCD02671629
CAS Number: 259153-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR