6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
Supplier: Matrix Scientific
010582-500MG
101798-800EA
15.38
USD
101798-800
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
Hazard: Irritant
Formula:
C₅H₆N₂O₃ MW: 142.12 g/mol Melting Pt: 195…196 °C |
MDL Number:
MFCD00052030 CAS Number: 27372-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...