1-(2-Fluorobenzyl)-1H-indole-3-carbaldehyde

Supplier: Matrix Scientific

010577-500MG
101798-792EA 267.69 USD
101798-792
1-(2-Fluorobenzyl)-1H-indole-3-carbaldehyde
1-(2-Fluorobenzyl)-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₂FNO
Melting Pt: 95…97 °C
MDL Number: MFCD02209555
CAS Number: 192997-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR